accessibility menu, dialog, popup

UserName

CAS RN 1569-08-0

IUPAC Name: 2-benzyl-4,4-dimethyl-4,5-dihydro-1,3-oxazole
Synonyms: 2-benzyl-4,4-dimethyl-5H-1,3-oxazole
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: GPBCTHQIXXYBHQ-UHFFFAOYSA-N
SMILES: CC1(C)COC(CC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results