accessibility menu, dialog, popup

UserName

CAS RN 154-17-6

IUPAC Name: 6-(hydroxymethyl)oxane-2,4,5-triol
Synonyms: 2-deoxy-D-glucose
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: PMMURAAUARKVCB-UHFFFAOYNA-N
SMILES: OCC1OC(O)CC(O)C1O
Quantity 
  • (4)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)