CAS RN 1539314-06-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GSK-2881078, MedChemExpress
MedChemExpress GSK 2881078 is a selective androgen receptor modulator potentially for the treatment of cachexia.