missing translation for 'onlineSavingsMsg'
Learn More

GSK-2881078, MedChemExpress

Product Code. 16499486
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499486 50 mg 50mg
16482828 10 mg 10mg
16425577 5 mg 5mg
16422536 25 mg 25mg
16410006 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499486 Supplier Medchem Express Supplier No. HY100186

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK 2881078 is a selective androgen receptor modulator potentially for the treatment of cachexia.

GSK 2881078 is a selective androgen receptor modulator potentially for the treatment of cachexia.

Specifications

Chemical Name or Material GSK-2881078
CAS 1539314-06-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H13F3N2O2S
Quantity 50 mg
Solubility Information DMSO : ≥ 33 mg/mL (99.90 mM)
SMILES N#CC1=C(C(F)(F)F)C2=C(N([C@H](C)CS(=O)(C)=O)C=C2)C=C1
Molecular Weight (g/mol) 330.33
Formula Weight 330.33
Percent Purity 99.74%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.