CAS RN 152273-12-6
IUPAC Name:
tert-butyl 4,4-dimethyl-4H,5H-imidazo[1,5-a]quinoxaline-3-carboxylate
Synonyms:
tert-butyl 4,4-dimethyl-5H-imidazo[1,5-a]quinoxaline-3-carboxylate
Molecular Weight (g/mol):
299.37
Molecular Formula:
C17H21N3O2
InChi Key:
NXBSEJKZKXIYMD-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)C1=C2N(C=N1)C1=CC=CC=C1NC2(C)C
1
–
1
of
1
results
1
U93631, MedChemExpress
MedChemExpress U93631 is a GABAA receptor ligand of novel chemical structure with IC50 of 100 nM,and has been shown to induce a rapid, time-dependent decay of GABA-induced whole-cell Cl-currents in recombinant GABAA receptors.