accessibility menu, dialog, popup

UserName

CAS RN 14930-96-2

IUPAC Name: 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
Synonyms: cytochalasen-B
Molecular Weight (g/mol): 479.62
Molecular Formula: C29H37NO5
InChi Key: GBOGMAARMMDZGR-UHFFFAOYNA-N
SMILES: CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (8)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (8)
  • (3)
  • (3)