accessibility menu, dialog, popup

UserName

FFFNH2NOO

CAS RN 148416-38-0

IUPAC Name: 2,3,4-trifluoro-6-nitroaniline
Synonyms: 2,3,4-trifluoro-6-nitroaniline
Molecular Weight (g/mol): 192.1
Molecular Formula: C6H3F3N2O2
InChi Key: ILENVKAVEFKZSD-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(F)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results