accessibility menu, dialog, popup

UserName

NHNNNHOOV

CAS RN 14705-63-6

IUPAC Name: vanadium(4+) 2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene dioxidandiide
Synonyms: vanadium(4+) tetraphenylporphyrin dioxidandiide
Molecular Weight (g/mol): 697.69
Molecular Formula: C44H30N4O2V
InChi Key: LRQXAIDMLNPKOR-UHFFFAOYSA-N
SMILES: [O--].[O--].[V+4].N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results