missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl Meso-Tetraphenylporphine, TRC

Product Code. 30450199
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450199 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450199 Supplier Toronto Research Chemicals Supplier No. TRCV96536510MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vanadyl Meso-Tetraphenylporphine
CAS 14705-63-6
Molecular Formula C44 H28 N4 O V
Quantity 10 mg
Synonym Vanadium','oxo[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-κN21,κN22,κN23,κN24]-','(SP-5-12)- (9CI','ACI), (SP-5-12)-Oxo[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-κN21,κN22,κN23,κN24]vanadium (ACI), 21H,23H-Porphine','5,10,15,20-tetraphenyl-','vanadium complex (ZCI), 21H,23H-Porphine','vanadium deriv. (ZCI), Porphine','tetraphenyl-','V complex (6CI), Vanadium','oxo[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-N21,N22,N23,N24]-','(SP-5-12)- (ZCI), Vanadium','oxo[5,10,15,20-tetraphenylporphinato(2-)]- (8CI), Vanadium','oxo[α,β,γ,Δ-tetraphenylporphinato(2-)]- (7CI), (5,10,15,20-Tetraphenyl-21H,23H-porphine)vanadium(IV) oxide, 5,10,15,20-tetraphenyl-21 H,23H-porphine vanadium(IV) oxide, Oxo(5,10,15,20-tetraphenylporphinato)vanadium, Oxo(tetraphenylporphyrinato)vanadium, Oxo[5,10,15,20-tetraphenylporphinato(2-)]vanadium, Oxovanadium(IV) tetraphenylporphyrin, Vanadyl 5,10,15,20-Tetraphenylporphyrin, Vanadyl tetraphenylporphine, Vanadyltetraphenylporphyrin
InChI Formula InChI=1S/C44H28N4.O.V/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;;/q-2;;+2/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;
SMILES O=[V+2]123N4=C5C=CC4=C(C6=CC=C(C(=C7C=CC(=N17)C(=C8C=CC(=C5c9ccccc9)[N-]28)c%10ccccc%10)c%11ccccc%11)[N-]36)c%12ccccc%12
Molecular Weight (g/mol) 679.66
Formula Weight 679.1703
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.