CAS RN 145689-34-5
IUPAC Name:
2-(2,3-difluorophenyl)acetonitrile
Synonyms:
2-(2,3-difluorophenyl)acetonitrile
Molecular Weight (g/mol):
153.13
Molecular Formula:
C8H5F2N
InChi Key:
IYRCHGRRMKOSHW-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(CC#N)=C1F
1
–
1
of
1
results
1
2,3-Difluorophenylacetonitrile, 97%, Thermo Scientific™
CAS: 145689-34-5 Molecular Formula: C8H5F2N Molecular Weight (g/mol): 153.13 MDL Number: MFCD00061277 InChI Key: IYRCHGRRMKOSHW-UHFFFAOYSA-N Synonym: 2,3-difluorophenylacetonitrile,2-2,3-difluorophenyl acetonitrile,2,3-difluorobenzyl cyanide,2,3-difluorophenylacetanitrile,benzeneacetonitrile, 2,3-difluoro,2-2,3-difluorophenyl ethanenitrile,2,3-difluorobenzeneacetonitrile,pubchem1587,acmc-209x0i,ksc183q4t PubChem CID: 518968 IUPAC Name: 2-(2,3-difluorophenyl)acetonitrile SMILES: FC1=CC=CC(CC#N)=C1F