CAS RN 14518-94-6
IUPAC Name:
cadmium(2+) dibromate
Synonyms:
cadmium(2+) dibromate
Molecular Weight (g/mol):
368.22
Molecular Formula:
Br2CdO6
InChi Key:
VECQEXBULIKSDM-UHFFFAOYSA-L
SMILES:
[Cd++].[O-][Br](=O)=O.[O-][Br](=O)=O
1
–
1
of
1
results
1
Cadmium bromate hydrate, 98%, Thermo Scientific™
CAS: 14518-94-6 Molecular Formula: Br2CdO6 Molecular Weight (g/mol): 368.22 MDL Number: MFCD04971194 InChI Key: VECQEXBULIKSDM-UHFFFAOYSA-L Synonym: cadmium bromate,cadmium dibromate,cadmium 2+ dibromate,2bro3.cd,acmc-1c8r8,bromic acid, cadmiumsalt 8ci,9ci PubChem CID: 14598111 SMILES: [Cd++].[O-][Br](=O)=O.[O-][Br](=O)=O