accessibility menu, dialog, popup

UserName

BOHNClClClHOOH

CAS RN 1449144-73-3

IUPAC Name: {2-chloro-5-[(2,3-dichlorophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-chloro-5-[(2,3-dichlorophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 344.38
Molecular Formula: C13H9BCl3NO3
InChi Key: JISRVYPGQJIELD-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(=C1)C(=O)NC1=C(Cl)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results