CAS RN 144032-36-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
rac N,O-Diacetyl Pseudoephedrine, TRC
CAS: 144032-36-0 Molecular Formula: C14 H19 N O3 Molecular Weight (g/mol): 249.31 Synonym: Acetamide, N-[2-(acetyloxy)-1-methyl-2-phenylethyl]-N-methyl-, (R*,R*)- (9CI),(1S,2S)-N,O-Diacetylpseudoephedrine,N-[(1S,2S)-2-(Acetyloxy)-1-methyl-2-phenylethyl]-N-methylacetamide IUPAC Name: [(1S,2S)-2-[acetyl(methyl)amino]-1-phenylpropyl] acetate SMILES: C[C@@H]([C@@H](OC(=O)C)c1ccccc1)N(C)C(=O)C
2
(1S,2S)-N,O-Diacetylpseudoephedrine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.