Learn More
(1S,2S)-N,O-Diacetylpseudoephedrine, Mikromol™
Product Code. 17941426
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17941426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17941426 Supplier Mikromol Supplier No. MM2115.05
 Restricted Item
Estimated Shipment: 25-09-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1S,2S)-N,O-Diacetylpseudoephedrine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1S,2S)-N,O-Diacetylpseudoephedrine
CAS 144032-36-0
Analyte or Component Names (1S,2S)-N,O-Diacetylpseudoephedrine
Shipping Condition Room Temperature
Molecular Formula C14 H19 N O3
Certifications/Compliance ISO 17025
Synonym (1S,2S)-N,O-Diacetylpseudoephedrine
InChI Formula InChI=1S/C14H19NO3/c1-10(15(4)11(2)16)14(18-12(3)17)13-8-6-5-7-9-13/h5-10,14H,1-4H3/t10-,14+/m0/s1
SMILES C[C@@H]([C@@H](OC(=O)C)c1ccccc1)N(C)C(=O)C
IUPAC Name [(1S,2S)-2-[acetyl(methyl)amino]-1-phenylpropyl] acetate
Molecular Weight (g/mol) 249.31
Quantity 100 mg
Formula Weight 249.1365 g/mol