accessibility menu, dialog, popup

UserName

ICH3P

CAS RN 14350-50-6

IUPAC Name: triphenyl(propyl)phosphanium iodide
Synonyms: triphenyl(propyl)phosphanium iodide
Molecular Weight (g/mol): 432.29
Molecular Formula: C21H22IP
InChi Key: MPNQDJZRGAOBPW-UHFFFAOYSA-M
SMILES: [I-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results