CAS RN 142994-45-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fmoc-d-3-pyridylalanine, TRC
CAS: 142994-45-4 Molecular Formula: C23H20N2O4 Molecular Weight (g/mol): 388.42 Synonym: (alphaR)-alpha-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-3-pyridinepropanoic Acid; SMILES: OC(=O)[C@@H](Cc1cccnc1)NC(=O)OCC2c3ccccc3c4ccccc24