accessibility menu, dialog, popup

UserName

H3CHN(R)CH3(S)(R)NH2(R)OO(R)(S)NH2(R)H2N(S)O(R)O(R)CH3HO(R)NHH3C(R)HO(S)OHCH3NH(R)(R)OH(R)O(R)H3COHO(S)(R)NH2(S)H2N(R)O(R)O(S)NH2(R)H2N(S)OHCH3NH(R)(R)OH(R)O(R)H3COHO(S)(R)NH2(S)H2N(R)O(R)O(S)(R)H2N(R)CH3NH2(S)OHOHSOHOO

CAS RN 1405-41-0

IUPAC Name: (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-(aminomethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(1R)-1-(methylamino)ethyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(1R)-1-aminoethyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; sulfuric acid
Synonyms: (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(1R)-1-aminoethyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; gentamicin C1; gentamicin C1A; sulfuric acid
Molecular Weight (g/mol): 1488.8
Molecular Formula: C60H125N15O25S
InChi Key: RDEIXVOBVLKYNT-HDZPSJEVSA-N
SMILES: OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](CN)CC[C@H]3N)[C@@H]2O)OC[C@]1(C)O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@@H](CC[C@H]3N)[C@@H](C)N)[C@@H]2O)OC[C@]1(C)O.CN[C@H](C)[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1
Quantity 
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)
  • (8)
  • (1)
Type 
  • (12)
Grade 
  • (1)
  • (3)
  • (5)
Color 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (3)
  • (5)
  • (10)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (8)
  • (1)
  • (12)
  • (1)
  • (3)
  • (5)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)