CAS RN 139680-47-0
IUPAC Name:
N,N-bis(2-methylpropyl)-4-nitrobenzamide
Synonyms:
N,N-bis(2-methylpropyl)-4-nitrobenzamide
Molecular Weight (g/mol):
278.35
Molecular Formula:
C15H22N2O3
InChi Key:
IFYJJQOERYQZBL-UHFFFAOYSA-N
SMILES:
CC(C)CN(CC(C)C)C(=O)C1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
N,N-Diisobutyl-4-nitrobenzamide, 97%, Thermo Scientific™
CAS: 139680-47-0 Molecular Formula: C15H22N2O3 Molecular Weight (g/mol): 278.352 MDL Number: MFCD01010614 InChI Key: IFYJJQOERYQZBL-UHFFFAOYSA-N Synonym: n,n-bis 2-methylpropyl-4-nitrobenzamide,n,n-diisobutyl-4-nitrobenzamide,benzamide, n,n-bis 2-methylpropyl-4-nitro,n,n-bis 2-methylpropyl 4-nitrophenyl carboxamide PubChem CID: 347941 IUPAC Name: N,N-bis(2-methylpropyl)-4-nitrobenzamide SMILES: CC(C)CN(CC(C)C)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]