CAS RN 1393-92-6
CAS RN 1393-92-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
6
of
6
results
1
Thermo Scientific Chemicals Litmus, pure, indicator grade
CAS: 1393-92-6 MDL Number: MFCD00131528
3
Litmus, MP Biomedicals™
CAS: 1393-92-6 MDL Number: MFCD00131528 Synonym: C.I. Natural Red 28,Lackmus,Tournesole,Chhadila
5
Litmus, Indicator (pH 5-8), for titration, Honeywell Fluka™
CAS: 1393-92-6 MDL Number: MFCD00000826
6
Litmus, Indicator, pH 5-8, for titration, Honeywell Fluka™
Indicator, pH 5-8, for titration