accessibility menu, dialog, popup

UserName

CAS RN 139256-79-4

IUPAC Name: 1-cyclopropylpiperazine dihydrochloride
Synonyms: 1-cyclopropylpiperazine dihydrochloride
Molecular Weight (g/mol): 199.12
Molecular Formula: C7H16Cl2N2
InChi Key: JDJNSFSTSJWJFA-UHFFFAOYSA-N
SMILES: Cl.Cl.C1CC1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results