CAS RN 1391737-01-1
CAS RN 1391737-01-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ VU 0463271
Potent and selective KCC2 inhibitor
2
VU0463271, MedChemExpress
MedChemExpress VU0463271 is a selective KCC2 antagonist, with an IC50 of 61 nM.
3
4