accessibility menu, dialog, popup

UserName

CAS RN 1373920-90-1

CH3OOHOFFFF

CAS RN 1373920-90-1

IUPAC Name: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid
Synonyms: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 238.14
Molecular Formula: C9H6F4O3
InChi Key: OVJKRERPBULYBW-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(=C1)C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results