CAS RN 1365267-36-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N1,N2-Bis[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-1,2-benzenedicarboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Bis-[Des(5-Chloro-2-carboxythienyl)] Rivaroxaban Phthalamide, TRC
CAS: 1365267-36-2 Molecular Formula: C36 H36 N6 O10 Molecular Weight (g/mol): 712.71 Synonym: N1,N2-bis[[(5S)-2-Oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-1,2-benzenedicarboxamide IUPAC Name: 1-N,2-N-bis[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]benzene-1,2-dicarboxamide SMILES: O=C(NC[C@H]1CN(C(=O)O1)c2ccc(cc2)N3CCOCC3=O)c4ccccc4C(=O)NC[C@H]5CN(C(=O)O5)c6ccc(cc6)N7CCOCC7=O