missing translation for 'onlineSavingsMsg'
Learn More

N1,N2-Bis[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-1,2-benzenedicarboxamide, Mikromol™

Product Code. 30051658
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051658 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051658 Supplier Mikromol Supplier No. MM3629.05
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N1,N2-Bis[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-1,2-benzenedicarboxamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N1,N2-bis[((5S)-2-Oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl)methyl]-1,2-benzenedicarboxamide
CAS 1365267-36-2
Analyte or Component Names N1,N2-bis[((5S)-2-Oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl)methyl]-1,2-benzenedicarboxamide
Shipping Condition Room Temperature
Molecular Formula C36 H36 N6 O10
Certifications/Compliance ISO 17025
Synonym N1,N2-bis[[(5S)-2-Oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-1,2-benzenedicarboxamide
InChI Formula InChI=1S/C36H36N6O10/c43-31-21-49-15-13-39(31)23-5-9-25(10-6-23)41-19-27(51-35(41)47)17-37-33(45)29-3-1-2-4-30(29)34(46)38-18-28-20-42(36(48)52-28)26-11-7-24(8-12-26)40-14-16-50-22-32(40)44/h1-12,27-28H,13-22H2,(H,37,45)(H,38,46)/t27-,28-/m0/s1
SMILES O=C(NC[C@H]1CN(C(=O)O1)c2ccc(cc2)N3CCOCC3=O)c4ccccc4C(=O)NC[C@H]5CN(C(=O)O5)c6ccc(cc6)N7CCOCC7=O
IUPAC Name 1-N,2-N-bis[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]benzene-1,2-dicarboxamide
Molecular Weight (g/mol) 712.71
Quantity 50 mg
Formula Weight 712.2493 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.