accessibility menu, dialog, popup

UserName

ClH3CNCH3(S)(S)(R)OHO(S)OHOH2NOHOOHNH3CCH3H(Z)

CAS RN 13614-98-7

IUPAC Name: hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Synonyms: hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-4a,5,5a,6-tetrahydro-4H-tetracene-1,3,12-trione chloride
Molecular Weight (g/mol): 493.94
Molecular Formula: C23H28ClN3O7
InChi Key: KDLQIOPKJDNQIM-WUURTAMISA-N
SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)