CAS RN 1317-39-1
CAS RN 1317-39-1
IUPAC Name:
bis(λ¹-copper(1+)) oxidandiuide
Synonyms:
cuprous oxide
Molecular Weight (g/mol):
145.11
Molecular Formula:
Cu2H2O
InChi Key:
FDARDWGARJUOKS-UHFFFAOYSA-N
SMILES:
[OH2--].[Cu+].[Cu+]
1
–
10
of
10
results
1
Copper(I) oxide, 97%
CAS: 1317-39-1 Molecular Formula: Cu2O Molecular Weight (g/mol): 143.09 MDL Number: MFCD00010974 Synonym: Cuprous oxide
2
Copper(I) oxide, 99% (metals basis)
CAS: 1317-39-1 Molecular Formula: Cu2O MDL Number: MFCD00010974
3
Copper(I) oxide, 99.9% (metals basis)
CAS: 1317-39-1 Molecular Formula: Cu2O MDL Number: MFCD00010974
4
Copper(I) oxide, 97% (Cu + Cu{2}O Assay)
CAS: 1317-39-1 Molecular Formula: Cu2O MDL Number: MFCD00010974
5
Copper(I) oxide, 97%
CAS: 1317-39-1 Molecular Formula: Cu2O MDL Number: MFCD00010974 Synonym: Cuprous oxide
6
Copper(I) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
7
Copper(I) Oxide, Honeywell Fluka™
CAS: 1317-39-1 Molecular Formula: Cu2O MDL Number: MFCD00010974
10
Copper(I) Oxide, 97%, Honeywell Fluka™
CAS: 1317-39-1 MDL Number: MFCD00010974