accessibility menu, dialog, popup

UserName

PtOO

CAS RN 1314-15-4

IUPAC Name: platinum(4+) dioxidandiide
Synonyms: adams's catalyst
Molecular Weight (g/mol): 227.08
Molecular Formula: O2Pt
InChi Key: WNVYCMIJBVIKSY-UHFFFAOYSA-N
SMILES: [O--].[O--].[Pt+4]
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (4)
  • (1)
  • (3)
Physical Form 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (4)
  • (1)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)