accessibility menu, dialog, popup

UserName

CAS RN 131395-23-8

IUPAC Name: 2-methoxy-4-(trifluoromethoxy)aniline
Synonyms: 2-methoxy-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 207.15
Molecular Formula: C8H8F3NO2
InChi Key: UHSZMPDNKZEQRZ-UHFFFAOYSA-N
SMILES: COC1=C(N)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results