accessibility menu, dialog, popup

UserName

CH3SNNNNCH3OOH3CH3CCH3ClCH3

CAS RN 1268524-70-4

IUPAC Name: tert-butyl 2-[7-(4-chlorophenyl)-4,5,13-trimethyl-3-thia-1,8,11,12-tetraazatricyclo[8.3.0.0²,⁶]trideca-2(6),4,7,10,12-pentaen-9-yl]acetate
Synonyms: tert-butyl 2-[7-(4-chlorophenyl)-4,5,13-trimethyl-3-thia-1,8,11,12-tetraazatricyclo[8.3.0.0²,⁶]trideca-2(6),4,7,10,12-pentaen-9-yl]acetate
Molecular Weight (g/mol): 456.99
Molecular Formula: C23H25ClN4O2S
InChi Key: DNVXATUJJDPFDM-UHFFFAOYNA-N
SMILES: CC1=C(C)C2=C(S1)N1C(C)=NN=C1C(CC(=O)OC(C)(C)C)N=C2C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results