accessibility menu, dialog, popup

UserName

CAS RN 12650-69-0

H3C(S)OH(S)CH3(S)O(S)(S)O(S)CH3OOOHO(R)HO(R)HO(E)

CAS RN 12650-69-0

IUPAC Name: 9-{[(2E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-{[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl}oxan-2-yl]-3-methylbut-2-enoyl]oxy}nonanoic acid
Synonyms: mupirocin mupirocin (substance) plasimine calcium salt (2:1) mupirocin mupirocin, 14c-labeled turixin MRC mupirocinum mupirocin (product) mupirocin, 14c labeled
Molecular Weight (g/mol): 500.63
Molecular Formula: C26H44O9
InChi Key: MINDHVHHQZYEEK-HBBNESRFSA-N
SMILES: C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]1CO[C@@H](C\C(C)=C\C(=O)OCCCCCCCCC(O)=O)[C@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (4)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results