missing translation for 'onlineSavingsMsg'
Learn More

Mupirocin, Mikromol™

Product Code. 17981466
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981466 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981466 Supplier Mikromol Supplier No. MM1340.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mupirocin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mupirocin
CAS 12650-69-0
Analyte or Component Names Mupirocin
Shipping Condition Ice pack (-20°C)
Molecular Formula C26 H44 O9
Certifications/Compliance ISO 17034
Synonym Mupirocin, 9-[[(2E)-4-[(2S,3R,4R,5S)-3,4-Dihydroxy-5-[[(2S,3S)-3-[(1S,2S)-2-hydroxy-1-methylpropyl]oxiranyl]methyl]tetrahydro-2H-pyran-2-yl]-3-methylbut-2-enoyl]oxy]nonanoic acid
InChI Formula InChI=1S/C26H44O9/c1-16(13-23(30)33-11-9-7-5-4-6-8-10-22(28)29)12-20-25(32)24(31)19(15-34-20)14-21-26(35-21)17(2)18(3)27/h13,17-21,24-27,31-32H,4-12,14-15H2,1-3H3,(H,28,29)/b16-13+/t17-,18-,19-,20-,21-,24+,25-,26-/m0/s1
SMILES C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]2CO[C@@H](C\C(=C\C(=O)OCCCCCCCCC(=O)O)\C)[C@H](O)[C@@H]2O
IUPAC Name 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
Molecular Weight (g/mol) 500.62
Quantity 250 mg
Formula Weight 500.2985 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.