accessibility menu, dialog, popup

UserName

CH3OFFFOH

CAS RN 1261747-88-9

IUPAC Name: 2-methyl-5-(trifluoromethoxy)phenol
Synonyms: 2-methyl-5-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 192.14
Molecular Formula: C8H7F3O2
InChi Key: IBICZXOZBCKSKR-UHFFFAOYSA-N
SMILES: CC1=C(O)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results