accessibility menu, dialog, popup

UserName

ClOFFFBr

CAS RN 1261606-06-7

IUPAC Name: 1-(bromomethyl)-4-chloro-2-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-4-chloro-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 289.48
Molecular Formula: C8H5BrClF3O
InChi Key: MJFOCXUYPQIXDL-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(CBr)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results