CAS RN 12612-37-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
9
of
9
results
1
AmberChrom 50Wx2 200-400 (H)
CAS: 12612-37-2 MDL Number: MFCD00132722
2
Amberchrom™ 50Wx2 100-200 (H)
CAS: 12612-37-2 MDL Number: MFCD00132722
3
Dowex™ 50WX2 50-100 (H)
CAS: 12612-37-2 MDL Number: MFCD00132722
4
Dowex 50X2-100, Ion-exchange Resin, ACROS Organics, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Dowex 50X2-100, Ion-exchange Resin, ACROS Organics, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
6
Dowex 50X2-100, Ion-exchange Resin, ACROS Organics, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
7
Dowex™ 50WX2, 100-200 mesh, ion-exchange resin
CAS: 12612-37-2 MDL Number: MFCD00132722
8
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
CAS: 12612-37-2 MDL Number: MFCD00132722
9
Dowex™ 50WX2, 50-100 mesh, ion-exchange resin
CAS: 12612-37-2 MDL Number: MFCD00132722