Learn More
Dowex 50X2-100, Ion-exchange Resin, ACROS Organics, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Dowex 50X2-100 |
| CAS | 12612-37-2 |
| Quantity | 1 g |
| Synonym | Ion-exchange Resin, ACROS Organics |