accessibility menu, dialog, popup

UserName

CAS RN 126-33-0

IUPAC Name: 1λ⁶-thiolane-1,1-dione
Synonyms: sulfolane
Molecular Weight (g/mol): 120.17
Molecular Formula: C4H8O2S
InChi Key: HXJUTPCZVOIRIF-UHFFFAOYSA-N
SMILES: O=S1(=O)CCCC1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (4)
  • (4)
  • (2)
  • (1)