accessibility menu, dialog, popup

UserName

SFFFFFClOOH(E/Z)

CAS RN 1240261-79-3

IUPAC Name: 3-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]prop-2-enoic acid
Synonyms: 3-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 308.65
Molecular Formula: C9H6ClF5O2S
InChi Key: ADQRQYHWENNZEP-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results