accessibility menu, dialog, popup

UserName

CAS RN 1214384-93-6

CH3OFFFOHO

CAS RN 1214384-93-6

IUPAC Name: 3-methoxy-2-(trifluoromethyl)benzoic acid
Synonyms: 3-methoxy-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: UOBKAUNYVVOXMY-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C(O)=O)=C1C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results