accessibility menu, dialog, popup

UserName

CH3OFFFO

CAS RN 1214384-89-0

IUPAC Name: 3-methoxy-2-(trifluoromethyl)benzaldehyde
Synonyms: 3-methoxy-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: HSPNVFXNJYKWSK-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results