CAS RN 1204144-28-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ AZD 1208
Potent pan Pim kinase inhibitor
2
AZD1208, TRC
CAS: 1204144-28-4 Molecular Formula: C21 H21 N3 O2 S Molecular Weight (g/mol): 379.48 Synonym: (5Z)-5-[[2-[(3R)-3-Amino-1-piperidinyl][1,1'-biphenyl]-3-yl]methylene]-2,4-thiazolidinedione IUPAC Name: (5Z)-5-[[2-[(3R)-3-aminopiperidin-1-yl]-3-phenylphenyl]methylidene]-1,3-thiazolidine-2,4-dione SMILES: N[C@@H]1CCCN(C1)c2c(\C=C\3/SC(=O)NC3=O)cccc2c4ccccc4
3
AZD1208, MedChemExpress
MedChemExpress AZD1208 is an orally bioavailable, highly selective PIM kinases inhibitor.