accessibility menu, dialog, popup

UserName

NH4I

CAS RN 12027-06-4

IUPAC Name: ammonium iodide
Synonyms: ammonium iodide ammoniumiodid
Molecular Weight (g/mol): 144.94
Molecular Formula: H4IN
InChi Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES: [NH4+].[I-]
Quantity 
  • (2)
  • (9)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (1)
Grade 
  • (2)
  • (7)
  • (2)
  • (3)
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (9)
  • (2)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (9)
  • (8)
  • (3)
  • (2)
  • (9)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (1)
  • (2)
  • (7)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (9)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (4)
  • (2)