accessibility menu, dialog, popup

UserName

CAS RN 12024-21-4

OOOGaGa

CAS RN 12024-21-4

IUPAC Name: digallium(3+) trioxidandiide
Synonyms: gallium(III) oxide
Molecular Weight (g/mol): 187.44
Molecular Formula: Ga2O3
InChi Key: AJNVQOSZGJRYEI-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Ga+3].[Ga+3]
Physical Form 
  • (2)
  • (12)
  • (3)
Quantity 
  • (1)
  • (3)
  • (5)
  • (6)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (7)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (15)
  • (1)
  • (2)
  • (12)
  • (3)
  • (1)
  • (3)
  • (5)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (7)
  • (3)
  • (2)