accessibility menu, dialog, popup

UserName

CAS RN 119-36-8

IUPAC Name: methyl 2-hydroxybenzoate
Synonyms: methyl salicylate methyl salicylate (product) betula caswell no. 577 metylester kyseliny salicylove sigma brand of methyl salicylate fema number 2745 gaultheriaoel hewedolor flucarmit
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: OSWPMRLSEDHDFF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (5)
  • (11)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (4)
Boiling Point 
  • (2)
  • (4)
  • (3)
Physical Form 
  • (6)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (11)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (4)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)