accessibility menu, dialog, popup

UserName

CAS RN 1180486-96-7

H3COClSFFFFF

CAS RN 1180486-96-7

IUPAC Name: 1-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]ethan-1-one
Synonyms: 1-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]ethanone
Molecular Weight (g/mol): 280.64
Molecular Formula: C8H6ClF5OS
InChi Key: MOKYPWIXBPTSMM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results