accessibility menu, dialog, popup

UserName

CAS RN 116795-47-2

IUPAC Name: 2-bromo-4-chloro-6-(hydroxymethyl)phenol
Synonyms: 2-bromo-4-chloro-6-(hydroxymethyl)phenol
Molecular Weight (g/mol): 237.48
Molecular Formula: C7H6BrClO2
InChi Key: RVRSNICKUNKOQP-UHFFFAOYSA-N
SMILES: OCC1=CC(Cl)=CC(Br)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results