CAS RN 116649-85-5
IUPAC Name:
3-[(3R)-3-(4-fluorobenzenesulfonamido)-2,3,4,9-tetrahydro-1H-carbazol-9-yl]propanoic acid
Synonyms:
ramatroban 雷马曲班 baynas
Molecular Weight (g/mol):
416.47
Molecular Formula:
C21H21FN2O4S
InChi Key:
LDXDSHIEDAPSSA-OAHLLOKOSA-N
SMILES:
OC(=O)CCN1C2=C(C[C@@H](CC2)NS(=O)(=O)C2=CC=C(F)C=C2)C2=CC=CC=C12
1
–
3
of
3
results
1
Ramatroban, TRC
CAS: 116649-85-5 Molecular Formula: C21 H21 F N2 O4 S Molecular Weight (g/mol): 416.47 Synonym: (3R)-3-[[(4-Fluorophenyl)sulfonyl]amino]-1,2,3,4-tetrahydro-9H-carbazole-9-propanoic Acid,BAY-u 3405,Baynas IUPAC Name: 3-[(3R)-3-[(4-fluorophenyl)sulfonylamino]-1,2,3,4-tetrahydrocarbazol-9-yl]propanoic acid SMILES: OC(=O)CCn1c2CC[C@H](Cc2c3ccccc13)NS(=O)(=O)c4ccc(F)cc4
2
Tocris Bioscience™ BAY-u 3405
CAS: 116649-85-5 Molecular Formula: C21H21FN2O4S
3
Ramatroban, MedChemExpress
MedChemExpress Ramatroban is a selective thromboxane A2 (TxA2, IC50=14 nM) antagonist, which also antagonizes CRTH2 (IC50=113 nM) by inhibiting PGD2 binding.