accessibility menu, dialog, popup

UserName

CAS RN 115619-00-6

H3CNNNOO

CAS RN 115619-00-6

IUPAC Name: 1-ethyl-4-(4-nitrophenyl)piperazine
Synonyms: 1-ethyl-4-(4-nitrophenyl)piperazine
Molecular Weight (g/mol): 235.29
Molecular Formula: C12H17N3O2
InChi Key: XEWICNRVCQLKIG-UHFFFAOYSA-N
SMILES: CCN1CCN(CC1)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results