CAS RN 1139-83-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Urolithin B, TRC
CAS: 1139-83-9 Molecular Formula: C13H8O3 Molecular Weight (g/mol): 212.2 Synonym: 3-Hydroxy-6H-dibenzo[b,d]pyran-6-one,2',4'-Dihydroxy-2-biphenylcarboxylic Acid δ-Lactone,3-Hydroxy-6H-benzo[c]chromen-6-one,3-Hydroxydibenzo-α-pyrone,3-Hydroxyurolithin,7-Hydroxy-3,4-benzocoumarin,NSC 94726 SMILES: Oc1ccc2c(OC(=O)c3ccccc23)c1