CAS RN 1120-06-5
CAS RN 1120-06-5
IUPAC Name:
decan-2-ol
Synonyms:
decan-2-ol
Molecular Weight (g/mol):
158.29
Molecular Formula:
C10H22O
InChi Key:
ACUZDYFTRHEKOS-UHFFFAOYNA-N
SMILES:
CCCCCCCCC(C)O
1
–
3
of
3
results
1
2-Decanol, 98%
CAS: 1120-06-5 Molecular Formula: C10H22O Molecular Weight (g/mol): 158.285 MDL Number: MFCD00004594 InChI Key: ACUZDYFTRHEKOS-UHFFFAOYSA-N Synonym: 2-decanol,2-hydroxydecane,2-decyl alcohol,methyl-n-octylcarbinol,decanol-2,methyl-n-octyl carbinol,acmc-1bovc,chembl46477 PubChem CID: 14254 IUPAC Name: decan-2-ol SMILES: CCCCCCCCC(C)O
2
2-Decanol, 98%
CAS: 1120-06-5 Molecular Formula: C10H22O Molecular Weight (g/mol): 158.28 MDL Number: MFCD00004594 InChI Key: ACUZDYFTRHEKOS-UHFFFAOYSA-N Synonym: 2-decanol,2-hydroxydecane,2-decyl alcohol,methyl-n-octylcarbinol,decanol-2,methyl-n-octyl carbinol,acmc-1bovc,chembl46477 PubChem CID: 14254 IUPAC Name: decan-2-ol SMILES: CCCCCCCCC(C)O