missing translation for 'onlineSavingsMsg'
Learn More

2-Decanol, MP Biomedicals

Product Code. 11448414 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448414 Supplier MP Biomedicals Supplier No. 0521085305
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1120-06-5
Molecular Formula C10H22O
Quantity 5 g
Synonym 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
InChI Key ACUZDYFTRHEKOS-UHFFFAOYSA-N
SMILES CCCCCCCCC(C)O
IUPAC Name decan-2-ol
Molecular Weight (g/mol) 158.285
PubChem CID 14254
Formula Weight 158.29
Chemical Name or Material 2-Decanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.